accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NONCH3NHNO

CAS RN 85872-53-3

IUPAC Name: 8-phenyl-1,3-dipropyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 8-phenyl-1,3-dipropyl-7H-purine-2,6-dione
Molecular Weight (g/mol): 312.37
Molecular Formula: C17H20N4O2
InChi Key: CLIGSMOZKDCDRZ-UHFFFAOYSA-N
SMILES: CCCN1C2=C(NC(=N2)C2=CC=CC=C2)C(=O)N(CCC)C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results