accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NOH

CAS RN 85951-60-6

IUPAC Name: 7-amino-5,6,7,8-tetrahydronaphthalen-2-ol
Synonyms: 7-amino-5,6,7,8-tetrahydronaphthalen-2-ol
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: VIYAPIMIOKKYNF-UHFFFAOYNA-N
SMILES: NC1CCC2=C(C1)C=C(O)C=C2
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results