accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCCH3CH3H3CCH3H3CCCH3CH3H3CCH3ClClHf

CAS RN 85959-83-7

IUPAC Name: hafnium(4+) bis(pentamethylcyclopenta-1,3-dien-1-ide) dichloride
Synonyms: hafnium(4+) bis(pentamethylcyclopenta-1,3-dien-1-ide) dichloride
Molecular Weight (g/mol): 519.85
Molecular Formula: C20H30Cl2Hf
InChi Key: YPRLSLCBPOTFGE-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Hf+4].CC1=[C-]C(C)(C)C(C)=C1C.CC1=[C-]C(C)(C)C(C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results