accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 859833-18-4

IUPAC Name: 1-[2-(pyridin-3-yl)phenyl]methanamine dihydrochloride
Synonyms: 1-[2-(pyridin-3-yl)phenyl]methanamine dihydrochloride
Molecular Weight (g/mol): 257.16
Molecular Formula: C12H14Cl2N2
InChi Key: SSFBEGXFELCUJT-UHFFFAOYSA-N
SMILES: Cl.Cl.NCC1=CC=CC=C1C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results