accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2S

CAS RN 859850-86-5

IUPAC Name: 1-[3-(thiophen-2-yl)phenyl]methanamine
Synonyms: 1-[3-(thiophen-2-yl)phenyl]methanamine
Molecular Weight (g/mol): 189.28
Molecular Formula: C11H11NS
InChi Key: PZXHYHVAVDOMOR-UHFFFAOYSA-N
SMILES: NCC1=CC=CC(=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results