CAS RN 86-28-2
CAS RN 86-28-2
IUPAC Name:
9-ethyl-9H-carbazole
Synonyms:
N-ethylcarbazol
Molecular Weight (g/mol):
195.27
Molecular Formula:
C14H13N
InChi Key:
PLAZXGNBGZYJSA-UHFFFAOYSA-N
SMILES:
CCN1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
9-Ethylcarbazole, 99%
CAS: 86-28-2 Molecular Formula: C14H13N Molecular Weight (g/mol): 195.27 MDL Number: MFCD00004967 InChI Key: PLAZXGNBGZYJSA-UHFFFAOYSA-N Synonym: n-ethylcarbazole,9-ethyl-9h-carbazole,9h-carbazole, 9-ethyl,carbazole, 9-ethyl,n-ethylcarbazol,n-ethyl carbazole,unii-6ak165l0ro,ccris 6847,9h-ethylcarbazole,9-ethyl carbazole PubChem CID: 6836 IUPAC Name: 9-ethylcarbazole SMILES: CCN1C2=C(C=CC=C2)C2=C1C=CC=C2