accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N

CAS RN 86-29-3

IUPAC Name: 2,2-diphenylacetonitrile
Synonyms: diphenylacetonitrile
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: NEBPTMCRLHKPOB-UHFFFAOYSA-N
SMILES: N#CC(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)