accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86-39-5

IUPAC Name: 2-chloro-9H-thioxanthen-9-one
Synonyms: 2-chlorothioxanthen-9-one
Molecular Weight (g/mol): 246.71
Molecular Formula: C13H7ClOS
InChi Key: ZCDADJXRUCOCJE-UHFFFAOYSA-N
SMILES: ClC1=CC=C2SC3=CC=CC=C3C(=O)C2=C1
Quantity 
  • (2)
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results