accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86-65-7

SOOHOSOOHOH2N

CAS RN 86-65-7

IUPAC Name: 7-aminonaphthalene-1,3-disulfonic acid
Synonyms: 7-aminonaphthalene-1,3-disulfonic acid
Molecular Weight (g/mol): 303.3
Molecular Formula: C10H9NO6S2
InChi Key: CMOLPZZVECHXKN-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=C(C=C(C2=C1)S(O)(=O)=O)S(O)(=O)=O
Grade 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)