accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HN

CAS RN 86-74-8

IUPAC Name: 9H-carbazole
Synonyms: 9H-carbazole diphenylenimide diphenylenimine diphenyleneimine usaf EK-600 dibenzopyrrole ccris 121 dibenzo(b,d)pyrrole
Molecular Weight (g/mol): 167.21
Molecular Formula: C12H9N
InChi Key: UJOBWOGCFQCDNV-UHFFFAOYSA-N
SMILES: N1C2=C(C=CC=C2)C2=C1C=CC=C2
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
Concentration 
  • (4)
  • (4)
  • (1)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (1)
  • (4)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)
  • (4)