CAS RN 86-81-7
CAS RN 86-81-7
IUPAC Name:
3,4,5-trimethoxybenzaldehyde
Synonyms:
3,4,5-trimethoxybenzaldehyde
Molecular Weight (g/mol):
196.2
Molecular Formula:
C10H12O4
InChi Key:
OPHQOIGEOHXOGX-UHFFFAOYSA-N
SMILES:
COC1=CC(C=O)=CC(OC)=C1OC
1
–
3
of
3
results
1
3,4,5-Trimethoxybenzaldehyde, 99%
CAS: 86-81-7 Molecular Formula: C10H12O4 Molecular Weight (g/mol): 196.2 MDL Number: MFCD00003364 InChI Key: OPHQOIGEOHXOGX-UHFFFAOYSA-N PubChem CID: 6858 IUPAC Name: 3,4,5-trimethoxybenzaldehyde SMILES: COC1=CC(=CC(=C1OC)OC)C=O
2
3,4,5-Trimethoxybenzaldehyde, 98%
CAS: 86-81-7 Molecular Formula: C10H12O4 Molecular Weight (g/mol): 196.202 MDL Number: MFCD00003364 InChI Key: OPHQOIGEOHXOGX-UHFFFAOYSA-N PubChem CID: 6858 IUPAC Name: 3,4,5-trimethoxybenzaldehyde SMILES: COC1=CC(=CC(=C1OC)OC)C=O
3
3,4,5-Trimethoxybenzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.