accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClN

CAS RN 86-98-6

IUPAC Name: 4,7-dichloroquinoline
Synonyms: quinoline, 4,7-dichloro-
Molecular Weight (g/mol): 198.05
Molecular Formula: C9H5Cl2N
InChi Key: HXEWMTXDBOQQKO-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(Cl)=CC=NC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (8)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (8)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)