accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNaOSOONHOHNOSOOO(E)

CAS RN 860-22-0

IUPAC Name: disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
Synonyms: indigo carmine 靛胭脂 acid blue 74
Molecular Weight (g/mol): 466.35
Molecular Formula: C16H8N2Na2O8S2
InChi Key: KHLVKKOJDHCJMG-QDBORUFSSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (4)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
Color 
  • (6)
  • (2)
  • (2)
Grade 
  • (3)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (6)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)