accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOSOCl

CAS RN 86010-32-4

IUPAC Name: 5-nitro-1-benzothiophene-2-carbonyl chloride
Synonyms: 5-nitro-1-benzothiophene-2-carbonyl chloride
Molecular Weight (g/mol): 241.65
Molecular Formula: C9H4ClNO3S
InChi Key: KOJDFXDLYGKDEK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2SC(=CC2=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results