accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NINI

CAS RN 86026-81-5

IUPAC Name: 2,5-diiodo-1-methyl-1H-imidazole
Synonyms: 2,5-diiodo-1-methylimidazole
Molecular Weight (g/mol): 333.9
Molecular Formula: C4H4I2N2
InChi Key: SZYKHAFVWJNSQU-UHFFFAOYSA-N
SMILES: CN1C(I)=CN=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results