accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNH2HCl

CAS RN 861036-67-1

IUPAC Name: 1-(quinolin-2-yl)methanamine hydrochloride
Synonyms: 1-(quinolin-2-yl)methanamine hydrochloride
Molecular Weight (g/mol): 194.66
Molecular Formula: C10H11ClN2
InChi Key: SUSYYDXUTFWSRV-UHFFFAOYSA-N
SMILES: Cl.NCC1=NC2=CC=CC=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results