accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNN

CAS RN 86236-77-3

IUPAC Name: 1,3-dibenzyl-1,3-diazinane-5-carbonitrile
Synonyms: 1,3-dibenzyl-1,3-diazinane-5-carbonitrile
Molecular Weight (g/mol): 291.4
Molecular Formula: C19H21N3
InChi Key: PSDQOOQJPHTEIA-UHFFFAOYSA-N
SMILES: N#CC1CN(CC2=CC=CC=C2)CN(CC2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results