accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86256-59-9

IUPAC Name: 2-methyl-4-(trifluoromethoxy)aniline
Synonyms: 2-methyl-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 191.15
Molecular Formula: C8H8F3NO
InChi Key: IIDBMILLZRYZCH-UHFFFAOYSA-N
SMILES: CC1=CC(OC(F)(F)F)=CC=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results