accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86267-86-9

IUPAC Name: 3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile
Synonyms: 3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: YSTANLOUKDVPGJ-UHFFFAOYNA-N
SMILES: N#CC1CNC2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results