CAS RN 86347-15-1
IUPAC Name:
hydrogen 5-[1-(2,3-dimethylphenyl)ethyl]-1H-imidazole chloride
Synonyms:
medetomidine
Molecular Weight (g/mol):
236.74
Molecular Formula:
C13H17ClN2
InChi Key:
VPNGEIHDPSLNMU-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CC(C1=CN=CN1)C1=CC=CC(C)=C1C
1
–
2
of
2
results
1
Medetomidine Hydrochloride, TRC
CAS: 86347-15-1 Molecular Formula: C13 H16 N2 . Cl H Molecular Weight (g/mol): 236.74 Synonym: Medetomidine Hydrochloride,1H-Imidazole, 5-[1-(2,3-dimethylphenyl)ethyl]-, hydrochloride (1:1),1H-Imidazole, 4-[1-(2,3-dimethylphenyl)ethyl]-, monohydrochloride (9CI),Domitor,MPV 785,Zalopine,5-[1-(2,3-Dimethylphenyl)ethyl]-1H-imidazole hydrochloride,4-(1-(2,3-Dimethylphenyl)ethyl)-1H-imidazole hydrochloride IUPAC Name: 4-[1-(2,3-dimethylphenyl)ethyl]-1H-imidazole;hydrochloride SMILES: Cl.CC(c1c[nH]cn1)c2cccc(C)c2C
2
Tocris Bioscience™ Medetomidine hydrochloride
CAS: 86347-15-1 Molecular Formula: C13H17ClN2