accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 863992-56-7

IUPAC Name: 4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 430.16
Molecular Formula: C26H32B2O4
InChi Key: ZLXSWNVYGSZXOP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results