accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86404-04-8

IUPAC Name: (5R)-5-[(1S)-1,2-dihydroxyethyl]-4-ethoxy-3-hydroxy-2,5-dihydrofuran-2-one
Synonyms: ethyl ascorbic acid 3-0-ethyl ascorbic acid 3-O-ethyl ascorbic acid
Molecular Weight (g/mol): 204.18
Molecular Formula: C8H12O6
InChi Key: ZGSCRDSBTNQPMS-UJURSFKZSA-N
SMILES: CCOC1=C(O)C(=O)O[C@@H]1[C@@H](O)CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results