CAS RN 864377-33-3
IUPAC Name:
[3-(9H-carbazol-9-yl)phenyl]boronic acid
Synonyms:
3-(carbazol-9-yl)phenylboronic acid
Molecular Weight (g/mol):
287.13
Molecular Formula:
C18H14BNO2
InChi Key:
IDQUIFLAFFZYEX-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(=CC=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
3-(9-Carbazolyl)benzeneboronic acid, 98%
CAS: 864377-33-3 Molecular Formula: C18H14BNO2 Molecular Weight (g/mol): 287.125 MDL Number: MFCD18072506 InChI Key: IDQUIFLAFFZYEX-UHFFFAOYSA-N Synonym: 3-9h-carbazol-9-yl phenylboronic acid,3-9h-carbazol-9-yl phenyl boronic acid,boronic acid, 3-9h-carbazol-9-yl phenyl,3-9-carbazolyl benzeneboronic acid,3-carbazol-9-yl phenylboronic acid,3-9h-9-carbozale phenylboronic acid,3-9h-9-carbazole phenylboronic acid,3-9h-carbazol-9-yl benzeneboronic acid PubChem CID: 56965778 IUPAC Name: (3-carbazol-9-ylphenyl)boronic acid SMILES: B(C1=CC(=CC=C1)N2C3=CC=CC=C3C4=CC=CC=C42)(O)O