accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3COCH3

CAS RN 864776-02-3

IUPAC Name: 4,4,5,5-tetramethyl-2-(2-methylfuran-3-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(2-methylfuran-3-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 208.06
Molecular Formula: C11H17BO3
InChi Key: JMLBPHNESOKSEV-UHFFFAOYSA-N
SMILES: CC1=C(C=CO1)B1OC(C)(C)C(C)(C)O1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results