accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86521-05-3

IUPAC Name: 2-[2-(trimethylsilyl)ethynyl]pyridine
Synonyms: 2-[2-(trimethylsilyl)ethynyl]pyridine
Molecular Weight (g/mol): 175.31
Molecular Formula: C10H13NSi
InChi Key: WOFPTESETJKCBH-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=CC=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results