accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86542-89-4

ONH2N

CAS RN 86542-89-4

IUPAC Name: 4-(azepane-1-carbonyl)piperidin-1-ium
Synonyms: 4-(azepane-1-carbonyl)piperidin-1-ium
Molecular Weight (g/mol): 211.33
Molecular Formula: C12H23N2O
InChi Key: BWKOHTHENZSVRL-UHFFFAOYSA-O
SMILES: O=C(C1CC[NH2+]CC1)N1CCCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results