accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86546-99-8

OOOOOOOOOEuEu

CAS RN 86546-99-8

IUPAC Name: dieuropium(3+) hydrate tricarbonate
Synonyms: dieuropium(3+) hydrate tricarbonate
Molecular Weight (g/mol): 501.97
Molecular Formula: C3H2Eu2O10
InChi Key: ZXWSECGKSXRERD-UHFFFAOYSA-H
SMILES: O.[Eu+3].[Eu+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results