accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86718-08-3

IUPAC Name: [4-(1H-imidazol-1-yl)phenyl]methanol
Synonyms: [4-(imidazol-1-yl)phenyl]methanol
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: SRQXVPAXMMIUFH-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)N1C=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results