accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

LiH3COCH3OH3COH3COH3COH3CONb

CAS RN 86745-52-0

IUPAC Name: niobium(5+) lithium(1+) hexaethanolate
Synonyms: niobium(5+) lithium(1+) hexakis(ethoxide)
Molecular Weight (g/mol): 370.21
Molecular Formula: C12H30LiNbO6
InChi Key: AVEKRHJBCCYCED-UHFFFAOYSA-N
SMILES: [Li+].[Nb+5].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)