accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOBrOOCH3

CAS RN 868-26-8

IUPAC Name: 1,3-dimethyl 2-bromopropanedioate
Synonyms: 1,3-dimethyl 2-bromopropanedioate
Molecular Weight (g/mol): 211.01
Molecular Formula: C5H7BrO4
InChi Key: NEMOJKROKMMQBQ-UHFFFAOYSA-N
SMILES: COC(=O)C(Br)C(=O)OC
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results