accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 86847-71-4

IUPAC Name: N-(3-formylpyridin-4-yl)-2,2-dimethylpropanamide
Synonyms: N-(3-formylpyridin-4-yl)-2,2-dimethylpropanamide
Molecular Weight (g/mol): 206.25
Molecular Formula: C11H14N2O2
InChi Key: ICMXCEJBHWHTBH-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)NC1=CC=NC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results