accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BNFFFHOOH

CAS RN 868662-36-6

IUPAC Name: [6-(trifluoromethyl)pyridin-3-yl]boronic acid
Synonyms: 6-(trifluoromethyl)pyridin-3-ylboronic acid
Molecular Weight (g/mol): 190.92
Molecular Formula: C6H5BF3NO2
InChi Key: BNTIPMNMTIAWIW-UHFFFAOYSA-N
SMILES: OB(O)C1=CN=C(C=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results