accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 868755-57-1

IUPAC Name: diphenyl-1,3-thiazole-5-sulfonyl chloride
Synonyms: diphenyl-1,3-thiazole-5-sulfonyl chloride
Molecular Weight (g/mol): 335.82
Molecular Formula: C15H10ClNO2S2
InChi Key: AWNJOBYLGNWNCQ-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results