accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 869-52-3

IUPAC Name: 3,6,9,12-tetrakis(carboxymethyl)-3,6,9,12-tetraazatetradecanedioic acid
Synonyms: 3,6,9,12-tetrakis(carboxymethyl)-3,6,9,12-tetraazatetradecanedioic acid
Molecular Weight (g/mol): 494.45
Molecular Formula: C18H30N4O12
InChi Key: RAEOEMDZDMCHJA-UHFFFAOYSA-N
SMILES: OC(=O)CN(CCN(CCN(CC(O)=O)CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results