accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNNONHNClH2COHOOOH

CAS RN 869497-75-6

IUPAC Name: 6-chloro-2-(4-methylpiperazine-1-carbonyl)-1H-1,3-benzodiazole; formic acid; prop-2-enoic acid
Synonyms: 5-chloro-2-(4-methylpiperazine-1-carbonyl)-3H-1,3-benzodiazole; acrylic acid; formic acid
Molecular Weight (g/mol): 396.83
Molecular Formula: C17H21ClN4O5
InChi Key: BODOIXYJOORQBQ-UHFFFAOYSA-N
SMILES: OC=O.OC(=O)C=C.CN1CCN(CC1)C(=O)C1=NC2=CC=C(Cl)C=C2N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results