accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 87-25-2

H3COOH2N

CAS RN 87-25-2

IUPAC Name: ethyl 2-aminobenzoate
Synonyms: ethyl anthranilate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: TWLLPUMZVVGILS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results