accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 87-28-5

IUPAC Name: 2-hydroxyethyl 2-hydroxybenzoate
Synonyms: glycol salicylate spirosal espirosal rheumacyl sarocol glysal glykolsalicylat kytta-gel traumasenex phlogont
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: LVYLCBNXHHHPSB-UHFFFAOYSA-N
SMILES: OCCOC(=O)C1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results