accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 87-32-1

IUPAC Name: (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
Synonyms: (2S)-2-acetamido-3-(1H-indol-3-yl)propanoate
Molecular Weight (g/mol): 245.26
Molecular Formula: C13H13N2O3
InChi Key: DZTHIGRZJZPRDV-LBPRGKRZSA-M
SMILES: CC(=O)N[C@@H](CC1=CNC2=CC=CC=C12)C([O-])=O
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)