accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 87-63-8

IUPAC Name: 2-chloro-6-methylaniline
Synonyms: 2-chloro-6-methyl-aniline
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: WFNLHDJJZSJARK-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Cl)=C1N
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)