accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ClHO

CAS RN 87-64-9

IUPAC Name: 2-chloro-6-methylphenol
Synonyms: 2-methyl-6-chlorophenol
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: YPNZJHFXFVLXSE-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Cl)=C1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)