accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 870-23-5

IUPAC Name: prop-2-ene-1-thiol
Synonyms: allyl mercaptan
Molecular Weight (g/mol): 74.14
Molecular Formula: C3H6S
InChi Key: ULIKDJVNUXNQHS-UHFFFAOYSA-N
SMILES: SCC=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)