accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 870-46-2

IUPAC Name: (tert-butoxy)carbohydrazide
Synonyms: tert-butoxycarbohydrazide
Molecular Weight (g/mol): 132.16
Molecular Formula: C5H12N2O2
InChi Key: DKACXUFSLUYRFU-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NN
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (6)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results