accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 870-72-4

IUPAC Name: sodium hydroxymethanesulfonate
Synonyms: formaldehyde sodium bisulfite
Molecular Weight (g/mol): 134.08
Molecular Formula: CH3NaO4S
InChi Key: UOULCEYHQNCFFH-UHFFFAOYSA-M
SMILES: [Na+].OCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)