CAS RN 87051-43-2
CAS RN 87051-43-2
IUPAC Name:
6-(2-{4-[bis(4-fluorophenyl)methylidene]piperidin-1-yl}ethyl)-7-methyl-5H-[1,3]thiazolo[3,2-a]pyrimidin-5-one
Synonyms:
ritanserin ritanserin tartrate tiserton 利坦色林 ritanserinum ritanserine ritanserina
Molecular Weight (g/mol):
477.57
Molecular Formula:
C27H25F2N3OS
InChi Key:
JUQLTPCYUFPYKE-UHFFFAOYSA-N
SMILES:
CC1=C(CCN2CCC(CC2)=C(C2=CC=C(F)C=C2)C2=CC=C(F)C=C2)C(=O)N2C=CSC2=N1
1
–
1
of
1
results
1