CAS RN 87100-28-5
IUPAC Name:
2-benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms:
2-benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol):
218.1
Molecular Formula:
C13H19BO2
InChi Key:
YCNQPAVKQPLZRS-UHFFFAOYSA-N
SMILES:
CC1(C)OB(CC2=CC=CC=C2)OC1(C)C
1
–
1
of
1
results
1
Benzylboronic acid pinacol ester, 96%
CAS: 87100-28-5 Molecular Formula: C13H19BO2 Molecular Weight (g/mol): 218.103 MDL Number: MFCD05663841 InChI Key: YCNQPAVKQPLZRS-UHFFFAOYSA-N Synonym: benzylboronic acid pinacol ester,1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl,acmc-209qek,benzylboronic pinacol ester,benzyl boronic acid pinacol ester,benzylboronic acid, pinacol ester,2-benzyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane,4,4,5,5-tetramethyl-2-phenylmethyl-1,3,2-dioxaborolane PubChem CID: 3864964 IUPAC Name: 2-benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC=C2