CAS RN 871126-15-7
IUPAC Name:
(3-fluoro-2-formylphenyl)boronic acid
Synonyms:
3-fluoro-2-formylphenylboronic acid
Molecular Weight (g/mol):
167.93
Molecular Formula:
C7H6BFO3
InChi Key:
GVHWLCYABLWGIR-UHFFFAOYSA-N
SMILES:
OB(O)C1=C(C=O)C(F)=CC=C1
1
–
1
of
1
results
1
3-Fluoro-2-formylbenzeneboronic acid, 95%
CAS: 871126-15-7 Molecular Formula: C7H6BFO3 Molecular Weight (g/mol): 167.93 MDL Number: MFCD10697421 InChI Key: GVHWLCYABLWGIR-UHFFFAOYSA-N Synonym: 3-fluoro-2-formylphenyl boronic acid,3-fluoro-2-formylbenzeneboronic acid,boronic acid, 3-fluoro-2-formylphenyl,acmc-209qf3,2-formyl-3-fluorophenylboranic acid,3-fluoro-2-formyl-phenyl boronic acid,3-fluoro-2-formylphenylboronic acid,boronic acid, b-3-fluoro-2-formylphenyl,contains varying amounts of anhydride PubChem CID: 53412038 IUPAC Name: (3-fluoro-2-formylphenyl)boronic acid SMILES: B(C1=C(C(=CC=C1)F)C=O)(O)O