accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 871329-62-3

BFOOCH3HOOH

CAS RN 871329-62-3

IUPAC Name: [3-fluoro-5-(methoxycarbonyl)phenyl]boronic acid
Synonyms: 3-fluoro-5-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 197.96
Molecular Formula: C8H8BFO4
InChi Key: VKHJVASTGLPBBL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results