accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 871366-38-0

BOOH3CCH3H3CH3CNOCH3(E/Z)

CAS RN 871366-38-0

IUPAC Name: N-(4-methoxyphenyl)-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanimine
Synonyms: N-(4-methoxyphenyl)-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanimine
Molecular Weight (g/mol): 337.23
Molecular Formula: C20H24BNO3
InChi Key: FEZZNCDZVGVBHZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N=CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results