accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HONNN

CAS RN 871825-54-6

IUPAC Name: {3-[(1H-1,2,4-triazol-1-yl)methyl]phenyl}methanol
Synonyms: [3-(1,2,4-triazol-1-ylmethyl)phenyl]methanol
Molecular Weight (g/mol): 189.22
Molecular Formula: C10H11N3O
InChi Key: PZRKSBSNVMSIJI-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(CN2C=NC=N2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results