CAS RN 872-50-4
1-Methyl-2-pyrrolidinone, 99%, extra pure
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.133 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N Synonym: 1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl PubChem CID: 13387 ChEBI: CHEBI:7307 IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.133 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N Synonym: 1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl PubChem CID: 13387 ChEBI: CHEBI:7307 IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.133 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N Synonym: 1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl PubChem CID: 13387 ChEBI: CHEBI:7307 IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, 99+%, ACS reagent
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.133 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N Synonym: 1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl PubChem CID: 13387 ChEBI: CHEBI:7307 IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, 99.5%, for HPLC
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.133 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N Synonym: 1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl PubChem CID: 13387 ChEBI: CHEBI:7307 IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.13 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, 99+%
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.13 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, HPLC Grade, 99.5%
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.13 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, 99+%, for spectroscopy
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.133 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N Synonym: 1-methyl-2-pyrrolidinone,n-methyl-2-pyrrolidone,n-methylpyrrolidone,1-methyl-2-pyrrolidone,methylpyrrolidone,m-pyrol,n-methyl-2-pyrrolidinone,n-methylpyrrolidinone,1-methylpyrrolidinone,2-pyrrolidinone, 1-methyl PubChem CID: 13387 ChEBI: CHEBI:7307 IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, ACS grade, 99.0+%
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.13 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.13 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
NMP, N-Methylpyrrolidone, GC Headspace Grade, Thermo Scientific™
1-Methyl-2-pyrrolidinone, 99.5%, for analysis
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.13 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, Spectrophotometric Grade, 99.5+%
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.13 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
1-Methyl-2-pyrrolidinone, Biograde, 99.5%
CAS: 872-50-4 Molecular Formula: C5H9NO Molecular Weight (g/mol): 99.13 MDL Number: MFCD00003193 InChI Key: SECXISVLQFMRJM-UHFFFAOYSA-N IUPAC Name: 1-methylpyrrolidin-2-one SMILES: CN1CCCC1=O
- 1
- 2